Single Factor Water Test Kits
Supplier: Industrial Test Systems
480125 487996-SFK 481108 481111 481110 480309 480104 481109
470144-534CS 678.05 CAD
470144-534 470144-546 470144-634 470173-830 470173-832 470173-838 470173-840 470173-834
Single Factor Water Test Kits
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Order Now
SPECIFICATIONS
Environmentally Preferable