Ward's® Simulated Blood Transfusion Matching Kit
Supplier: Avantor
470023-682KT 101.25 CAD
470023-682 470023-688
Ward's® Simulated Blood Transfusion Matching Kit
Educational Classroom Kits and Activities
In complete safety, this kit teaches students how to determine blood types.
  • Pre- and Post-Laboratory Assessments Included
  • Kit Introduces Tests Essential to Successful Blood Transfusions
  • Tests Link to Real-World Concepts and Professional Practices
  • Two-Part Lab, 30 Minutes Each
  • Material for Six Setups
They perform the same tests hematologists use. Students also practice cross-matching, which is vital for successful blood transfusions.
Students use Ward's Simulated Blood to re-create ABO blood-typing techniques practiced in real labs and determine what types are compatible for transfusions. This kit includes a teacher’s guide, a student copymaster, and enough material for six setups.
Ordering information: This kit includes Simulated Blood of Recipient: Type B; Simulated Blood of Mr. Smith: Type A; Simulated Blood of Mr. Jones: Type B; Simulated Blood of Mr. Green: Type AB; Simulated Blood of Ms. Brown: Type O, simulated anti-A typing serum, simulated anti-A typing serum, simulated recipient serum, blood-typing slides, a package of toothpicks, and micro-centrifuge tubes. A micro-centrifuge is required but not included.