Simple Cuboidal Epithelium Slides
Supplier: Avantor
470177-562EA 12.55 CAD
470177-562 470182-610 470183-660
Simple Cuboidal Epithelium Slides
Slides Prepared Slides
Aid in secretion and absorption of organs.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Choose from several available stains and preparations for these tissue sections to get the view you're looking for. Slides are made from mammal kidney, and demonstrate cuboidal cells in tubules. Thin section slide is plastic-embedded for greater viewing clarity.