Ward's® Live Silkworm Eggs (Bombyx mori)
Supplier: COASTAL SILKWORMS
SWE2000 DF-1/4
470180-380PK 39.4 CAD
470180-380 470180-382
Ward's® Live Silkworm Eggs (Bombyx mori)
Specimens Live Specimens Animals - Living Material
When hatched, the small, gray larvae are the traditional source for high-quality silk.
  • They can remain refrigerated for up to a year, allowing for easy storage and access
  • Eggs will hatch in 10 - 14 days if kept at 75 - 85 °F
Over thousands of years of domestication and selective breeding the modern silkworm has lost its ability to fly. Eggs, each roughly the size of a pinhead, come glued onto a petri dish.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2ndMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable