Silica Gel

Supplier: ALDON CORP SE

SS0150-500G SS0150-500GI
470302-360EA 80.8 CAD
470302-360 470302-362
Silica Gel
Chemicals for Science Education

CAS Number: 112926-00-8
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 2.1
Boiling Point (°C): 2230
Freezing Point (°C): 1710
Synonyms: Synthetic Amorphous Silica Gel, Indicating
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR