Sharp-Point Surgical Scissors
Supplier: Dunrite Instruments
140982 140981 140980
470018-942BX 146.45 CAD
470018-942 470018-940 470106-340
Sharp-Point Surgical Scissors
Scissors
Start your cuts easily.
  • Dissect without a scalpel
  • Handles rounded for ambidextrous use
  • Available in two sizes
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable