Sharpening Stone
Supplier: REXEL USA
NOR61463687565
470150-112EA 75.6 CAD
470150-112
Sharpening Stone
Scissors
Prolong the life of your important dissecting instruments by sharpening them often on this ultrafine grit sharpening stone (Arkansas novaculite).
Size: 3"L x 1"W