Ward's® Shale (Argillaceous)
Supplier: Avantor
470026-256PK 26.2 CAD
470026-256 470026-258 470026-260
Ward's® Shale (Argillaceous)
Specimens Rock Specimens Sedimentary Rocks
Typical green-gray, fissile, clay-rich.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram pack can be cutMore Product Information