Shake, Rattle & Roll Earthquake Board
Supplier: Avantor
470016-770EA 614.75 CAD
470016-770 470039-398
Shake, Rattle & Roll Earthquake Board
Educational Classroom Kits and Activities
This wave maker earthquake board and kit makes seismic waves in the classroom.
  • Teacher-Developed and Tested
  • Reusable
  • For Grades Six through Nine
  • Teacher's Guide Included
  • Board Makes Seismic Waves in the Classroom
  • Side-to-Side Movement
The first part of this complete unit describes how to use a Slinky® spring to demonstrate four types of seismic waves: primary, secondary, Raleigh, and L-waves. Students then place their Towering Toothpick structures onto the board and create “quakes” of gradually increasing magnitude to test the durability of theirMore Product Information