Seines
Supplier: DOUGLAS NET COMPANY
N2018WHITE P2518BROWN
470106-846EA 191 CAD
470106-846 470106-354
Seines
Animal Supplies Animal Nets
Great for Stream Sampling
  • Large Area Screen for Sampling
  • Collect More Fish and Specimens
Suitable for both saltwater and freshwater, this seine features 2 mm mesh and includes weights and floats. Size: 1m x 7.6m.