Screw Top Dropper Pipet Bottle
Supplier: United Scientific Supplies
BDPSC-1OZ BDPSC-0.5OZ BDPSC-2OZ BDPSC-1OZ
470012-952PK 94.7 CAD
470012-952 470325-644 470325-658 470325-660
Screw Top Dropper Pipet Bottle
Bottles Dropping Bottles
Molded of flint glass, screw tip with reusable, snap-on sealer tip.
  • Glass pipet
  • Closure included
  • 30 ml capacity
These Boston round flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving.
Packaging: Pack of 12