Screw Caps for Specimen Jars
Supplier: EMPIRE/EMCO
87A001D89 87A001I58
470175-872EA 2.95 CAD
470175-872 470189-036 470189-038
Screw Caps for Specimen Jars
Closures Bottle Closures
These steel caps fit flint glass, screw cap wide mouth specimen jars. They are finished in black enamel and have a chemically resistant inner liner.