Vermiculite Spill Clean Up Material

Supplier: ALDON CORP SE
VV0025-1GAL VV0025-1KG
470303-008EA 47 CAD
470303-008 470303-010
Vermiculite Spill Clean Up Material
Spill Control Materials
CAS Number: 1318-00-9
Formula Weight: Mixture
Formula: Mixture
Density (g/ml): 2.5
Freezing Point (°C): 1350
Solubility: insoluble
Shelf Life (months): 36
Storage: Green

For 470303-008 there is 400g fill of vermiculite within 1 gallon container

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR