Ward's® Chemistry Instant Light in the Bag Inquiry Activity
Supplier: Avantor
LL0255-100G
470016-838EA 77.1 CAD
470016-838 470301-302
Ward's® Chemistry Instant Light in the Bag Inquiry Activity
Educational Classroom Kits and Activities
With this activity, teachers just add water to a dry powder for an intriguing, instant illumination in a bag.
  • Activity Demonstrates Chemiluminescence
  • Quick, Easy, Mess-Free Demonstration
  • Kit Includes 10 Heavy-Duty Bags and 100 g of Instant Light Crystals
  • Ten Heavy-Duty Bags Provided
The mixture glows in the dark, and students discover the chemistry behind chemiluminescence. They explore it by designing experiments to make the light brighter or last longer.
Students begin to understand that the light is a product, and the kinetics of the reaction can be controlled by temperature. The activity is a great introduction to kinetics, scientific inquiry, and chemiluminescence.
Ordering information: This kit includes 100 g bottle of instant light crystals, instructions, and 10 heavy duty plastic bags.