Ward's® Chemistry Ferrofluid Demonstration
Supplier: EDUCATIONAL INNOVATIONS, INC.
WARD470016-854 FF-200 AD-FF0195-50ML WARD470016-852
470093-856EA 239.3 CAD
470093-856 470217-236 470016-854 470016-852
Ward's® Chemistry Ferrofluid Demonstration
Educational Classroom Kits and Activities
Amaze and Excite Your Students!
  • Ideal for teaching magnetism, forces and chemistry, Grades 9-12
  • Great demonstration for younger students, Grades 5-8
  • Learn about nanomaterials and their properties
  • Investigate technical applications for ferrofluids
Amaze and excite your students with ferrofluid, an incredible substance which is in liquid form until a strong magnetic field is brought near it. As the magnet comes closer to the ferrofluid, spikes begin to rise from it. Ask students to determine the reason for this peculiar reaction. Ferrofluid, a colloid with magnetic nanoparticles suspended in an oil base, was originally discovered by NASA scientists. Now ferrofluids are used to make rotating seals in high vacuum equipment and can be even found in high end speakers.
Ordering information: Materials include Vinyl Gloves, Jar with Lid, Instructions, Petri Dish, Ferrofluid, Glass Vials with Caps, Ceramic Block Magnet, Cylindrical Magnet, Nylon String, Plastic Jar with Lid, Modeling Clay, Test Tubes with Stoppers, and Bar Magnet. Pennies, Scotchgard™ spray, Water, Paper towels, Liquid dish soap, Lab coat/apron, Safety glasses are materials needed but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.