Scheibler Desiccators
Supplier: Variety Glass
682-3 682-5 682-3 470204-328
25010-060EA 224.2 CAD
25010-060 25010-122 470204-324 470204-328
Scheibler Desiccators
Desiccators Desiccators, Glass
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly. Porcelain plates sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable