Ward's® Live Sarcina aurantiaca Culture
Supplier: VWR
WARD470176-648 WARD470176-646
470176-648EA 55.2 CAD
470176-648 470176-646
Ward's® Live Sarcina aurantiaca Culture
Specimens Live Specimens Bacteria - Living Material
Produces orange pigments. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Cocci in tetrads and packets
  • Gram Reaction: Positive
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.