Sand

Supplier: ALDON CORP SE

SS0080-1KG SS0090-2.5KG SS0090-500G SS0060-5KG SS0090-5KG SS0060-500G SS0080-500G
470302-338EA 24.4 CAD
470302-338 470302-340 470302-344 470302-334 470302-342 470302-336 470302-346
Sand
Chemicals for Science Education

CAS Number: 14808-60-7
Formula: SiO2
Density: 2.65 g/mL
Boiling and Freezing Point: 2230°C, 1610°C
Solubility: Insoluble
Synonyms: Silicon Dioxide, Quartz, Crystalline Silica
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR