Ward's® Live Salmonella typhimurium, Ames Test Strain Culture - PATHOGEN
Supplier: Avantor
470179-146EA 20.45 CAD
470179-146 470179-580
Ward's® Live Salmonella typhimurium, Ames Test Strain Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Ames test strain for detecting carcinogens. Causes infection in humans. 
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need toMore Product Information