Salicylic Acid
Supplier: ALDON CORP SE
CAS Number: 69-72-7
Formula Weight: 138.12
Formula: C7H6O3
Density (g/mL): 1.443
Boiling Point (°C): 211
Freezing Point (°C): 158-160
Solubility: Water and Organic Solvents
Synonyms: 2-Hydroxybenzoic acid
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...