Saf-T-Scalpels
Supplier: DR INSTRUMENTS INC. SE
140911 4-510 4-515
470014-846EA 13.05 CAD
470014-846 470203-414 470203-416
Saf-T-Scalpels
Scalpels and Scalpel Assemblies Scalpels
Safer solution for your students.
  • Retractable blade for safety
  • Disposable
  • Plastic handle allows for no slip operation
Easily operate these retractable blade scalpels with simple movement of the thumb on one hand. Each scalpel features an integrated double safety feature to prevent accidental blade extension and injury during dissection procedures. These quality instruments are color-coded, cost-effective, and compliant with OSHA requirements. Non sterile scalpels are designed for educational dissections.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.