Safranin O

Supplier: ALDON CORP SE

SS0030-500ML SS0030-100ML SS0020-10G SS0020-25GR SS0029-30ML
470302-320EA 62.9 CAD
470302-320 470302-318 470302-322 470302-324 470302-316
Safranin O
Chemicals for Science Education

CAS Number: 477-73-6
Formula: C20H19N4Cl
Density: 1 g/mL
Solubility: Water and Alcohol
Synonyms: Basic Red 2, C.I. No. 50240
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR