SAFE-T Compass
Supplier: LEARNING RESOURCES SE
45760 45760
470201-062PK 74.15 CAD
470201-062 470046-182
SAFE-T Compass
Compasses
Perform all standard functions accurately without dangerous points.
  • Durable plastic
  • Straight edges
Compass can draw circles 1/4” to 10” diameter in 1/16” radius increments. Direct view of vertex points allows for easier and more accurate constructions. Straight edge sides calibrated with inch/metric rules.
Ordering information: Includes complete instructions.