Saccharomyces cerevisiae
Supplier: VWR
470179-656EA 22.05 CAD
470179-656
Saccharomyces cerevisiae
Specimens Live Specimens Fungi - Living Material
Ascomycete. Host for rDNA. Incubation temperature 30°C. Sabouraud dextrose/YM agar growth medium.