Saccharomyces cerevisiae
Supplier: CONNECTICUT VALLEY BIO SUPPLY
L9040P
470176-658EA 22.05 CAD
470176-658 470177-272
Saccharomyces cerevisiae
Specimens Live Specimens Fungi - Living Material
Ascomycete. Baker’s yeast. Reproduction by budding. Incubation temperature 30°C. YM agar growth medium.