Rubber Laboratory Apron
Supplier: VWR International
470148-648
470148-648CS 694.95 CAD
470148-648
Rubber Laboratory Apron
Workwear
Durable, lightweight, rubber apron resistant to chemicals, acids, and caustics.
  • Chemical-resistant
  • Extra-heavy rubber material
  • Lightweight
  • Contains latex
Extra-heavy rubber coated sheeting plus cotton tabs on the waist and neck have made these aprons a durable lab standard for years. They resist most chemicals, acids, and caustics, yet are still lightweight.