PYREX® Narrow Mouth Reagent Bottles, Corning Life Sciences
Supplier: Corning
1500-250 1500-250
470200-306CS 3069.75 CAD
470200-306 470211-376
PYREX® Narrow Mouth Reagent Bottles, Corning Life Sciences
Bottles Glass Bottles
PYREX® borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers. Replacement stoppers are available.