Round Carboys
Supplier: Thermo Fisher Scientific
2318-0010 2318-0020
470211-768CS 1407.35 CAD
470211-768 470211-770
Round Carboys
Bottles Carboys
Suitable for collecting and dispensing distilled water, reagents, and acids, these low-density polyethylene carboys are graduated in 1 gallon and 5 L increments.
  • Two larger sizes also have convenient wide shoulder handles
The polypropylene spigot is molded in one piece with the bottle for added strength and leak resistance.