Ward's® Rhyolite (Pink)
Supplier: Avantor
470005-180EA 27.95 CAD
470005-180 470015-496 470026-184
Ward's® Rhyolite (Pink)
Specimens Rock Specimens Igneous Rocks
Pale pink felsite, with small sanidine phenocrysts.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram packMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable