Rescue Critters® K-9 Thoracentesis Manikin
Supplier: Rescue Critters
925
470126-196EA 5532.79 CAD
470126-196
Rescue Critters® K-9 Thoracentesis Manikin
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Emergency Response Training For Vets
  • Convenient anatomical landmarks
  • Perform chest tube placements
  • Made to order
This carefully crafted canine Thoracentesis manikin will help practitioners in performing chest tube placements, and simulating emergency trauma by aspirating air and fluid from the thoracic cavity. Convenient anatomical landmarks are placed at key points for ease of use and quick learning.
Ordering information: This item is made to order. Returns cannot be accepted.