Rectangular Glass Coverslips
Supplier: C & A Scientific
94-2240 94-2440 94-22402
470019-008PK 11.7 CAD
470019-008 470019-010 470145-746
Rectangular Glass Coverslips
Cover Glasses
Traditional rectangular coverslip style
  • Quality slide making supplies
  • Traditional coverslip type
Traditional style coverslips to use when making slides in the laboratory.
#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.