Rectangular Bases
Supplier: Wards
S78311-10C WLS78311-10A WLS78311-10D
470016-124EA 39.95 CAD
470016-124 470123-470 470123-474
Rectangular Bases
Retort Stands
Essential for Any Chemistry or Physics Laboratory
  • 3 mm thick steel
  • Baked-on enamel finish
  • 2 tapped hole positions
One hole is in the base top at the center of the shorter side for standard ring-stand use and one other at the center of the longer side for titrations and specific gravity determinations.
Ordering information: Rod sold separately.