Rapitest Soil Test Kit
Supplier: LUSTER LEAF PRODUCTS INC SE
1612 1601
470104-846CS 156.35 CAD
470104-846 470175-090
Rapitest Soil Test Kit
Standards Environmental Analysis Standards
Perform qualitative soil tests without the use of chemical reagents and test tubes.
  • No hazardous compounds required for testing
  • Colorimetric determination of soil components
  • Instructions included
This kit lets students use a simple three-step process involving water, tablets and color comparators. To test soil, simply mix it with water and let it settle. Then pour the liquid off into the color comparator, add a tablet and compare the liquid with the color film chart for a test reading.
470104-846 includes 10 pH soil test kits and fully instructions.
470175-090 includes 10 tests each for pH, nitrogen, phosphorus and potash and full instructions.