Boreal Science Quieting Solution
Supplier: ALDON CORP SE
BB0161-100 BB0161-30M
470007-320EA 19.5 CAD
470007-320 470106-046
Boreal Science Quieting Solution
Chemicals for Science Education
Use this solution to slow down and relax microorganisms such as Paramecium, Euglena and Stentor. The ready-to-use dropper bottle makes it easy to store and transfer quieting liquid. Includes a dropper.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.