PYREX® Erlenmeyer flasks, Wide Neck, Heavy-Duty Rim
Supplier: Corning
5100-125 5100-1L 5100-250 5100-500
470211-494CS 809.25 CAD
470211-494 470211-496 470211-498 470211-500
PYREX® Erlenmeyer flasks, Wide Neck, Heavy-Duty Rim
Flasks Erlenmeyer Flasks
Heavy-duty rim flasks are especially recommended for use as titration flasks.
  • White enamel graduation to show the approximate capacity
  • With extra large marking spot
  • Good thermal and chemical resistance
Designed for long life and maximum safety, these flasks have a wide mouth, as indicated by stopper openings.