PYREX® Standard-Grade Beakers
Supplier: Corning
1000-10 1000-20 1000-2L 1000-30 1000-800 1000-150
470211-308CS 536.1 CAD
470211-308 470211-318 470211-322 470211-324 470211-334 470211-312
PYREX® Standard-Grade Beakers
Beakers
Made of PYREX® borosilicate glass.
  • PYREX®, Soda-lime glass, with spout
  • Very good chemical resistance
  • Good temperature resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
Certifications: Meet ASTM E960 specifications.
Ordering information: The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.