United Scientific™ Pull-Type Spring Scales (Metric/Newton)
Supplier: United Scientific Supplies
SB0250-P SB0500-P SB2000-P SB3000-P SBTSET6 SB5000-P
470000-122EA 14.4 CAD
470000-122 470000-124 470020-002 470020-004 470339-336 470339-970 470349-726
United Scientific™ Pull-Type Spring Scales (Metric/Newton)
Balances and Scales
With these scales, students see the spring action as it occurs.
  • Color-coded
  • Clear plastic tube
  • Zero adjustment
These color-coded spring scales have a clear-tube design that allows students to see the spring action as it occurs. Each scale is 7 inches long, and features zero adjustment. The set of six comes with one of each weight capacity.
Caution: Caution:WARNING: CHOKING HAZARD This product contains small parts is not for children under three years.