Pulley Cords
Supplier: Arcor Electronics
SDX16165 6CB-15 10PB-135 50CD-130 30LB50YARD
470005-414EA 31 CAD
470005-414 470014-396 470110-156 470110-158 470105-858 470201-332
Pulley Cords
Educational Materials Physics Educational Materials Mechanics Learning Activities
Ideal for small pulley systems, cord is available in a variety of colors, strengths, and lengths.
  • Ideal for pulley systems, hanging masses, and wave demonstrations
  • Nylon and Dacron are extremely durable
  • Ends may be heat fused to prevent fraying
  • Premium Dacron cords resists stretching and fraying
Every physics classroom needs good-quality string - whether it's for an Atwood's machine, pulling friction blocks, or hanging pendulums. Teachers need string that is easy to knot and won't unravel after it is cut. These products have been selected for durability and strength, and come on a convenient spool for easy dispensing and storage.