Ward's® Live Pseudomonas putida Culture
Supplier: VWR
WARD470179-092
470179-092EA 22.05 CAD
470179-092
Ward's® Live Pseudomonas putida Culture
Specimens Live Specimens Bacteria - Living Material
From soil and water rich in minerals. Organism of putrefaction.
  • Biosafety Level: 1
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.