Ward's® Live Proteus mirabilis Culture - PATHOGEN
Supplier: Avantor
470179-138EA 22.05 CAD
470179-138 470179-536
Ward's® Live Proteus mirabilis Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Isolated from sewage, human intestines, and urinary tract infections. Spreading colonies are seen on agar growth medium. 
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-More Product Information