Probe And Seeker
Supplier: Dunrite Instruments
140958 WLS26907
470018-924EA 8.85 CAD
470018-924 470122-006
Probe And Seeker
Dissection Needles
This tool performs double duty, with a curved probe at one end and a flat spade at the other to gently separate muscles without tearing or cutting. Stainless steel.
Order Now
SPECIFICATIONS
Environmentally Preferable