Equilateral Acrylic Prisms
Supplier: United Scientific Supplies
PAE025 PAE075 PAE150 PAE100
470004-204EA 19.35 CAD
470004-204 470013-002 470101-252 470101-250
Equilateral Acrylic Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate refraction and internal reflection.
  • Highly polished sides
  • High optical clarity
Selection of equilaterial and right angle acrylic prisms.
Order Now
SPECIFICATIONS
Environmentally Preferable