Prepared Electrophoresis Agar
Supplier: ALDON CORP SE
AA84-200ML
AA0079-200
DEFAULTVPN
470191-730EA
27
CAD
470191-730
470191-732
470191-734
Prepared Electrophoresis Agar
Media for Microbiology
Prepared Media for Microbiology
Jump to the electrophoresis step quicker - let us prepare your gel for you.
- Convenient, just melt and pour.
- Choose the concentration to view DNA fragments from 0.1 to 35 kb in length
2.0% Agarose is used to resolve DNA fragments in the range of 0.1 to 5kb in length.
0.8% Agarose is used to resolve DNA fragments in the range of 0.6-35kb in length.