Precision Snow/Rain Gauge
Supplier: SCIENTIFIC SALES INC SE
6331
470146-182EA 122.79 CAD
470146-182
Precision Snow/Rain Gauge
Weather Stations
Measures in metric units.
  • Can be attached to the included rust–proof mounting bracket
  • Aligned with GLOBE protocols
This gauge, made of clear butyrate, has an internal measuring tube with a capacity of 1" (25 mm) and is graduated to 0.01" (0.2 mm). Any precipitation greater than 1" overflows into the outer cylinder and is measured by pouring the overflow into the measuring tube. The receiving funnel and measuring tube can be removed for snow collection. It includes a sturdy, rust–proof mounting bracket, and is available in metric units.