Precision pH Test Strips
Supplier: Precision Laboratories
PH0114-1B-50 PH3060-1B-50 PH1013-1B-50 PH5090-1B-50 PH1114-1B-50
470001-956PK 15.8 CAD
470001-956 470150-922 470150-806 470150-846 470150-880 470150-918 470151-004
Precision pH Test Strips
Test Papers pH Indicator Papers and Test Strips
The indicator on these strips won’t contaminate the solutions the strips are testing.
  • Vinyl-backed
  • Non-contaminating indicator
  • Fifty strips per package
Durable pH testing strips that are backed with vinyl.