Potassium Thiocyanate

Supplier: ALDON CORP SE

PP0801-500ML PP0802-CONC PP0790-100G PP0790-500G PP0790-500GR
470302-276EA 17.15 CAD
470302-276 470302-278 470302-280 470302-282 470302-286
Potassium Thiocyanate
Chemicals for Science Education

CAS Number: 333-20-0
Formula Weight: 97.18
Formula: KSCN
Hazard Info: Toxic
Density (g/mL): 1.886
Boiling Point (°C): 500
Freezing Point (°C): 173
Solubility: Water, Alcohol, and Acetone
Synonyms: Thiocyanic Acid Potassium Salt, Potassium Sulfocyanate, Potassium Rhodanide
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR