Potassium Nitrite
Supplier: ALDON CORP SE
CAS Number: 7758-09-0
Formula Weight: 85.11
Formula: KNO2
Hazard Info: Oxidizer, Flammable, Irritant
Density (g/mL): 1.915
Boiling Point (°C): Explodes at 538
Solubility: Water
Synonyms: Nitrous Acid, Potassium Salt
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...