Potassium Nitrate

Supplier: ALDON CORP SE

PP0650-2.5KG PP0650-500G PP0655-500GR PP0650-CAP PP0667-500ML PP0662-CONC PP0650-100G PP0655-100GR PP0655-2.5KGR
470302-194EA 112.4 CAD
470302-194 470302-198 470302-204 470302-188 470302-190 470302-192 470302-196 470302-200 470302-202
Potassium Nitrate
Chemicals for Science Education

CAS Number: 7757-79-1
Formula: KNO3
Density: 2.1 g/mL
Boiling and Freezing Point: 400°C, 333°C
Solubility: Water, Glycerin, and Alcohol
Synonyms: Potash Nitrate, Saltpeter
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable oxidizer and irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR