Potassium Iodide

Supplier: ALDON CORP SE

PP0629-500ML PP0610-500G PP0610-100GR PP0610-25GR PP0610-100G PP0630-1L PP0630-500G PP0629-CONC PP0610-25G PP0610-500GR PP0606-500ML PP0626-500ML
470302-166EA 99.85 CAD
470302-166 470302-174 470302-176 470302-178 470302-170 470302-162 470302-164 470302-168 470302-172 470302-180 470225-734 470225-788
Potassium Iodide
Chemicals for Science Education

CAS Number: 7681-11-0
Formula: KI
Density: 3.12 g/mL
Boiling and Freezing Point: 1330°C, 680°C
Solubility: Water, Glycerin, Alcohol, and Acetone
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR