Potassium Iodate

Supplier: ALDON CORP SE

PP0601-100G PP0601-100GR PP0603-500ML PP0601-500G PP0601-25GR PP0601-500GR PP0605-500ML PP0605-1L
470302-152EA 72.3 CAD
470302-152 470302-156 470302-150 470302-154 470302-158 470302-160 470225-648 470225-786
Potassium Iodate
Chemicals for Science Education

CAS Number: 7758-05-6
Formula Weight: 214.00
Formula: KIO3
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.98
Solubility: Water and Dilute Sulfuric Acid
Synonyms: Iodic Acid, Potassium Salt
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR